PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,5 ml, Violet
Katalog # 731-4031
Leverandør: Corning
Gå til:
Product Details & Documents
Polypropylene tubes are specifically designed for PCR and thermal transfer applications.
- Made from highly polished molds
- Free of lubricants, dyes, heavy metals, and fillers
- Certified RNase-, DNase-, and pyrogen-free to ensure the purity of your reaction
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling.
Specifikationer
- Farve:Violet
- Beskrivelse:PCR microtubes with flat cap, 0,5 ml
- Seq:07