Orders LinkContactUsLinkComponent
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,5 ml, blå
Katalog # 731-4026
Leverandør:   Corning
PCR microtubes with flat cap, Axygen®
Katalog # 731-4026
Leverandør:  Corning
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,5 ml, blå
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Polypropylene tubes are specifically designed for PCR and thermal transfer applications.
  • Made from highly polished molds
  • Free of lubricants, dyes, heavy metals, and fillers
  • Certified RNase-, DNase-, and pyrogen-free to ensure the purity of your reaction
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling.
Specifikationer
  • Farve:
    blå
  • Beskrivelse:
    PCR microtubes with flat cap, 0,5 ml
  • Seq:
    06