PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,2 ml, gul
731-4005
: Corning
Polypropylene tubes are specifically designed for PCR and thermal transfer applications.
- Made from highly polished molds
- Free of lubricants, dyes, heavy metals, and fillers
- Certified RNase-, DNase-, and pyrogen-free to ensure the purity of your reaction
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling.
- Farve:gul
- Beskrivelse:PCR microtubes with flat cap, 0,2 ml
- Seq:05
- Cat. No.:731-4005
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Lav fremstillingspåvirkning