Orders LinkContactUsLinkComponent
Pipettespidser, universal, Axygen®
Pipettespidser, universal, Axygen®
Bevelled, Maxymum Recovery®, 15 - 1250 µl, -, Transparent, 12 racks, 96 each
Katalog # 613-1265
Pipettespidser, universal, Axygen®
Katalog # 613-1265
Leverandør:  
Pipettespidser, universal, Axygen®
Pipettespidser, universal, Axygen®
Bevelled, Maxymum Recovery®, 15 - 1250 µl, -, Transparent, 12 racks, 96 each
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
PIPETTESPIDS 1250UL, MATRIX, FILTER, STE 1 * 1.152 Items Katalog #613-1269
Produktet 613-1269 ligner 613-1265, men producenten og specifikationerne kan variere.
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Bestillingsoplysninger: Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
  • Pakket:
    12 racks, 96 each
  • Steril:
    -
  • Volumen:
    15 - 1250 µl
  • Seq:
    71
  • Spidstype:
    Bevelled, Maxymum Recovery®
  • Spits farge:
    Transparent