Orders LinkContactUsLinkComponent
Pipettespidser, universal, Axygen®
Bevelled, Matrix-style, 15 - 1250 µl, -, Transparent, Bulk
Katalog # 613-1263
Leverandør:   Corning
Pipettespidser, universal, Axygen®
Katalog # 613-1263
Leverandør:  Corning
Pipettespidser, universal, Axygen®
Pipettespidser, universal, Axygen®
Bevelled, Matrix-style, 15 - 1250 µl, -, Transparent, Bulk
Price
479,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
 : Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
  • Pakket:
    Bulk
  • Steril:
    -
  • Volumen:
    15 - 1250 µl
  • Seq:
    68
  • Spidstype:
    Bevelled, Matrix-style
  • Spits farge:
    Transparent