Pipettespidser, universal, Axygen®
Bevelled, Matrix-style, 15 - 1250 µl, -, Transparent, Bulk
Katalog # 613-1263
Leverandør: Corning
Gå til:
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
- Pakket:Bulk
- Steril:-
- Volumen:15 - 1250 µl
- Seq:68
- Spidstype:Bevelled, Matrix-style
- Spits farge:Transparent