Pycnometers, unadjusted
5 ml
Catalog # WITG3.900.005
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in capillary stopper
- Nominal capacity printed on the bottom
- Very good chemical and temperature resistance
Certifications: DIN 12797
Specifications
- Volumen (nom.):5 ml