Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
E.Z.N.A.® Plasmid Midi kit

E.Z.N.A.® Plasmid Midi kit

E.Z.N.A.® Plasmid Midi Kit I, 100 Tests
Katalog # D6904-04
Leverandør:   OMEGA BIO-TEK
E.Z.N.A.® Plasmid Midi kit
Katalog # D6904-04
Leverandør:  OMEGA BIO-TEK
CAS nummer:  
E.Z.N.A.® Plasmid Midi kit
E.Z.N.A.® Plasmid Midi kit

E.Z.N.A.® Plasmid Midi Kit I, 100 Tests

CAS nummer:  
Price
6.500,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

E.Z.N.A.® Plasmid Midi Kit offers a novel and reliable method of isolating up to 200 µg of plasmid DNA in less than 60 minutes. This kit combines HiBind® membrane technology with the time-tested consistency of alkaline SDS lysis of bacterial cells to deliver high-quality plasmid DNA. The DNase/RNase-free HiBind® DNA midi columns operate by eliminating time consuming phenol-chloroform extractions and alcohol precipitations. Plasmid purification using the E.Z.N.A.® Plasmid Midi Kit follows a simple bind-wash-elute procedure thereby making it possible for multiple samples to be processed. Although yields vary according to plasmid copy number, E.coli strain, and conditions of growth, 50 ml of an overnight culture in LB medium will typically produce 200 µg of high-copy number plasmid DNA. When working with low-copy number plasmids the starting culture volume can be increased up to 100 ml. High-quality DNA is ready for immediate use in routine molecular biology applications, such as automated fluorescent sequencing, restriction enzyme digestion, and transfection screening.

  • Purification of plasmid DNA in <60 minutes
  • No phenol/chloroform extractions
  • DNA suitable for most downstream applications
Specifikationer

Specifikationer

  • Pakn:
    100 Tests
  • Beskrivelse:
    E.Z.N.A.® Plasmid Midi Kit I
  • Cat. No.:
    D6904-04
  • Prep size:
    Midi
  • Beskrivelse:
    E.Z.N.A.® Plasmid Midi Kit I
  • Refine target molecule:
    Plasmid
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Bacteria
  • Sample size:
    15 - 50 ml

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.