Ribonuclease A
Bottle (2,5 ml), 2,5 mL
Leverandør: OMEGA BIO-TEK
Gå til:
Product Details & Documents
- Enzyme activity is 100 mg/ml (for 5 ml)
Documents
Specifikationer
Specifikationer
- Pakning:Bottle (2,5 ml)
- Pakn:2,5 mL
- Cat. No.:NA-03
- CAS-nr:9001-99-4
- MDL No.:MFCD00132184
- Opbevaring betingelser:Freezer
- Enzym navn:Ribonuclease A
- Enzym synonymer:RNase A
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



