Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Urglas

VWR®, Urglas

Watch glass, PTFE, 75 mm
Katalog # 216-0159
Leverandør:   VWR Collection
VWR®, Urglas
Katalog # 216-0159
Leverandør:  VWR Collection
VWR®, Urglas
VWR®, Urglas

Watch glass, PTFE, 75 mm

Price
93,30 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

PTFE.

  • Excellent chemical resistance
  • Temperature resistant from –200 to 280 °C
  • Exceptionally smooth surfaces to prevent contamination

Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.

Specifikationer
  • Ø:
    75 mm
  • Beskrivelse:
    Watch glass, PTFE
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.