Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Centrifugerør

50 ml, 25×145 mm, uden cap
  525-0181
VWR®, Centrifugerør
  525-0181
VWR®, Centrifugerør
VWR®, Centrifugerør

50 ml, 25×145 mm, uden cap

 
285,00 kr
   
1 * 1 Items
 

PTFE, rund bund, glat overflade, uden skruelåg.

  • Inert
  • Withstand forces up to 12500×g
  • Excellent chemical resistance

Egnet til brug ved høje og lave temperaturer (–200 til +280 °C).

 
  • Lukkestørrelse:
    uden cap
  • Ø×L:
    25×145 mm
  • Kapacitet:
    50 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.