Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Urglas

Watch glass, PTFE, 150 mm
  216-0163
VWR®, Urglas
  216-0163
VWR®, Urglas
VWR®, Urglas

Watch glass, PTFE, 150 mm

 
240,00 kr
   
1 * 1 Items
 

PTFE.

  • Excellent chemical resistance
  • Temperature resistant from –200 to 280 °C
  • Exceptionally smooth surfaces to prevent contamination

Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.

 
  • Ø:
    150 mm
  • Beskrivelse:
    Watch glass, PTFE