Closures for test tubes and flasks
Vatkugler, store, ikke-sterile
Katalog # 734-1265
Leverandør: US COTTON
Gå til:
Product Details & Documents
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
Specifikationer
- Beskrivelse:Vatkugler, store, ikke-sterile
- Seq:09