Orders LinkContactUsLinkComponent
Closures for test tubes and flasks
Closures for test tubes and flasks
Vatkugler, store, ikke-sterile
Katalog # 734-1265
Leverandør:   US COTTON
Closures for test tubes and flasks
Katalog # 734-1265
Leverandør:  US COTTON
Closures for test tubes and flasks
Closures for test tubes and flasks
Vatkugler, store, ikke-sterile
Price
906,38 kr
Log ind for tilgængelighed
1 * 2.000 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
Specifikationer
  • Beskrivelse:
    Vatkugler, store, ikke-sterile
  • Seq:
    09