Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
Cover glasses, square, without cords and blisters, 25×50 mm, #1
  KNITVD12550Y100A
 :   KNITTEL
Cover glasses
  KNITVD12550Y100A
 :  KNITTEL
Cover glasses
Cover glasses, square, without cords and blisters, 25×50 mm, #1
1.210,00 kr
1 * 1.000 Items
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certifications: ISO 8255/1
Packaging: 100 pieces per box - 1000 pieces per carton.
Documents
  • Antal:
    #1
  • Beskrivelse:
    Cover glasses, square, without cords and blisters
  • L×B:
    25×50 mm