Cover glasses
Cover glasses, square, without cords and blisters, 25×50 mm, #1
KNITVD12550Y100A
: KNITTEL
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certifications: ISO 8255/1
Packaging: 100 pieces per box - 1000 pieces per carton.
Documents
KNITVD11818Y100A_TDS_EN
Datasheet
- Antal:#1
- Beskrivelse:Cover glasses, square, without cords and blisters
- L×B:25×50 mm