Orders LinkContactUsLinkComponent
Accessories for syringes, SampleLock™
Accessories for syringes, SampleLock™
Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
Katalog # HAMI35081
Leverandør:   HAMILTON BONADUZ
Accessories for syringes, SampleLock™
Katalog # HAMI35081
Leverandør:  HAMILTON BONADUZ
Accessories for syringes, SampleLock™
Accessories for syringes, SampleLock™
Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
Price
457,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These are a series of adapters that convert a large hub removable needle (RN termination) syringe to a Luer connection. There are three different removable adapters for 250 µl and larger syringes.
The adapters convert the RN termination into either a Male Luer, Male Luer Lock or a Female Luer Lock connection. The adapter has a port inner diameter of 0,99 mm that allows for sample spiking through a syringe fitted with a 26 gauge needle or smaller.
Specifikationer
  • Beskrivelse:
    Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
  • Seq:
    02