Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tilbehør til pH-elektroder

Tilbehør til pH-elektroder

Sensor cable VP 6, max. Ø 7,5 mm, length 5 m
Catalog # 662-2083
Supplier:   HAMILTON BONADUZ
Tilbehør til pH-elektroder
Catalog # 662-2083
Supplier:  HAMILTON BONADUZ
Tilbehør til pH-elektroder

Sensor cable VP 6, max. Ø 7,5 mm, length 5 m

Price
2.220,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Sensor cable VP 6, max. Ø 7,5 mm, length 5 m
  • Seq:
    16
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.