Orders LinkContactUsLinkComponent
Ficoll® PM70
Ficoll® PM70
Ficoll® PM70
Katalog # 17-0310-05
Ficoll® PM70
Katalog # 17-0310-05
Leverandør:  
Ficoll® PM70
Ficoll® PM70
Ficoll® PM70
Discontinued
Product Details & Documents
Ficoll® PM70 is a high molecular weight sucrose polymer formed by copolymerisation of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
  • Displays minimal toxicity
  • Favourable viscosity and osmotic properties
The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
Specifikationer
  • Beskrivelse:
    Ficoll® PM70