5,5'-Dithiobis(2-nitrobenzosyre)
100 Tests
CAYM400050-100
: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- :69-78-3
- :C₁₄H₈N₂O₈S₂
- :MFCD00007140
- :396.36 g/mol
- :240…244 °C
- :Stue temperatur
- :200-714-4
- Pakn:100 Tests