Orders LinkContactUsLinkComponent
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
Decapper til 20 mm krympelåg
Catalog # WHEAW225353
Supplier:   DWK Life Sciences
Decappers, E-Z Decappers™, WHEATON®
Catalog # WHEAW225353
Supplier:  DWK Life Sciences
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
Decapper til 20 mm krympelåg
Price
7.090,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Autoclavable
Specifications
  • Beskrivelse:
    Decapper til 20 mm krympelåg
  • Seq:
    04