Decappers, E-Z Decappers™, WHEATON®
Decapper til 11 mm krympelåg
Katalog # WHEAW225351
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
- Cushioned ergonomic handle reduces hand fatigue
- Labeled for quick size identification
- Autoclavable
Specifikationer
- Beskrivelse:Decapper til 11 mm krympelåg
- Seq:02