Orders LinkContactUsLinkComponent
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
Decapper til 8 mm krympelåg
Katalog # WHEAW225350
Leverandør:   DWK Life Sciences
Decappers, E-Z Decappers™, WHEATON®
Katalog # WHEAW225350
Leverandør:  DWK Life Sciences
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
Decapper til 8 mm krympelåg
Price
9.960,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Autoclavable
Specifikationer
  • Beskrivelse:
    Decapper til 8 mm krympelåg
  • Seq:
    01