Eco caps
Replacement male Luer plug, PP
Katalog # JOURJR-070350-10
Leverandør: VICI JOUR RESEARCH
Gå til:
Product Details & Documents
The Eco Cap consists of a three-hole ETFE insert with a polyethylene collar.
- Compatible with all standard GL 45 thread bottles
- Also available as bottle kit
There is no threading for fittings on the inside of the tubing ports, the tubing can be easily inserted.
Leveringsinformation: The Eco Cap is supplied with one cap, polypropylene male Luer plug. The kit also includes safety-coated glass bottle with a option of 1 or 2 L.
Specifikationer
- Beskrivelse:Replacement male Luer plug, PP
- Seq:04