Orders LinkContactUsLinkComponent
Nuts and ferrules
Nuts and ferrules
PPS hex-head nut, for flange/washer connections, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
Catalog # JOURJR-55061-10
Supplier:   VICI JOUR RESEARCH
Nuts and ferrules
Catalog # JOURJR-55061-10
Supplier:  VICI JOUR RESEARCH
Nuts and ferrules
PPS hex-head nut, for flange/washer connections, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
Price
367,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    PPS hex-head nut, for flange/washer connections, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
  • Seq:
    10