Nuts and ferrules
PPS hex-head nut, flangeless, for use with inverted ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
Catalog # JOURJR-55070-10
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:PPS hex-head nut, flangeless, for use with inverted ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
- Seq:07