Orders LinkContactUsLinkComponent
Nuts and ferrules
Nuts and ferrules
PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
Katalog # JOURJR-55050-10
Leverandør:   VICI JOUR RESEARCH
Nuts and ferrules
Katalog # JOURJR-55050-10
Leverandør:  VICI JOUR RESEARCH
Nuts and ferrules
PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
Price
351,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
  • Seq:
    01