Nuts and ferrules
PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
Katalog # JOURJR-55050-10
Leverandør: VICI JOUR RESEARCH
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/16" O.Ø tubing or 1/4"–28 flat bottom ports
- Seq:01