Nuts and ferrules
PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
JOURJR-55051-10
- Beskrivelse:PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
- Seq:02