Orders LinkContactUsLinkComponent
Nuts and ferrules
PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
  JOURJR-55051-10
Nuts and ferrules
  JOURJR-55051-10
Nuts and ferrules
PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
351,00 kr
1 * 10 Items
  • Beskrivelse:
    PPS hex-head nut, flangeless, for use with collapsible ferrule, for 1/8" O.Ø tubing or 1/4"–28 flat bottom ports
  • Seq:
    02