PCR microtubes without caps, 0,2 ml, Amplitube™
ufarvet, 0,2 ml
SIMPT325-12N
Made of PP, translucent.
- Choice of colorless and four non-cytotoxic and non-metallic colors
- Very good chemical resistance
- Very good temperature resistance
This thin wall 0,2 ml tube is very useful when processing smaller volumes. It offers optimum contact with thermal cycler blocks. The ultrathin wall will ensure rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Specially designed with a highly polished surface and a round bottom for maximum sample recovery.
: Caps must be ordered seperately.
: Packed in tamperproof resealable bags.
- Kapacitet:0,2 ml
- Farve:ufarvet