Orders LinkContactUsLinkComponent
PCR microtubes without caps, 0,2 ml, Amplitube™
PCR microtubes without caps, 0,2 ml, Amplitube™
blå, 0,2 ml
Katalog # SIMPT325-12B
Leverandør:   Simport Scientific
PCR microtubes without caps, 0,2 ml, Amplitube™
Katalog # SIMPT325-12B
Leverandør:  Simport Scientific
PCR microtubes without caps, 0,2 ml, Amplitube™
PCR microtubes without caps, 0,2 ml, Amplitube™
blå, 0,2 ml
Price
476,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Made of PP, translucent.
  • Choice of colorless and four non-cytotoxic and non-metallic colors
  • Very good chemical resistance
  • Very good temperature resistance
This thin wall 0,2 ml tube is very useful when processing smaller volumes. It offers optimum contact with thermal cycler blocks. The ultrathin wall will ensure rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Specially designed with a highly polished surface and a round bottom for maximum sample recovery.
Ordering information: Caps must be ordered seperately.
Packaging: Packed in tamperproof resealable bags.
Specifikationer
  • Kapacitet:
    0,2 ml
  • Farve:
    blå