Orders LinkContactUsLinkComponent
Forceps
Forceps
Fine point, straight, 120 mm
Katalog # 241-0338
Leverandør:   EXCELTA
Forceps
Katalog # 241-0338
Leverandør:  EXCELTA
Forceps
Fine point, straight, 120 mm
Price
511,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel, antimagnetic.
  • Economical special forceps for precision applications
  • Corrosion-resistant
Ideal when working with microelectronic assemblies and in the electronics industry.
Specifikationer
  • Beskrivelse:
    Fine point, straight
  • Længde:
    120 mm
  • Seq:
    06