Orders LinkContactUsLinkComponent
Screwthread joints for VACUU.LAN®
Screwthread joints for VACUU.LAN®
Straight adapter, 45 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
Katalog # VACU20638778
Leverandør:   VACUUBRAND
Screwthread joints for VACUU.LAN®
Katalog # VACU20638778
Leverandør:  VACUUBRAND
Screwthread joints for VACUU.LAN®
Straight adapter, 45 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
Price
61,50 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Adapters for VACUU.LAN® local area vacuum network components.
Specifikationer
  • Beskrivelse:
    Straight adapter, 45 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
  • Seq:
    06