Orders LinkContactUsLinkComponent
HPLC columns, MonoChrom® C18
HPLC columns, MonoChrom® C18
Forkolonne, C18, 110 Å, 5 µm
Katalog # VARIA0505MG
Leverandør:   VARIAN
HPLC columns, MonoChrom® C18
Katalog # VARIA0505MG
Leverandør:  VARIAN
HPLC columns, MonoChrom® C18
Forkolonne, C18, 110 Å, 5 µm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 3 Items
Product Details & Documents
MonoChrom® columns are ideal for separation of organic amines, organic acids, amphoteric compounds and neutral compounds. Advanced silica production and silane bonding techniques allow for the production of material with exceptional base-deactivated characteristics.
Specifikationer
  • Kolonne form:
    Forkolonne
  • Partikelstørrelse:
    5 µm
  • Porestørrelse:
    110 Å
  • Seq:
    14
  • Fase:
    C18