Slangekoblinger, Y-formede
12/13/14 mm, 7,6 mm, 12 - 14 mm
: KARTELL
PP, hvid.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Suitable for foodstuff
Med koniske ender.
Certifications: Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520.
- Udv.Ø:12/13/14 mm
- Hul Ø:7,6 mm
- Rør int.Ø:12 - 14 mm
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



