Slangekoblinger, Y-formede
10/11/12 mm, 5,5 mm, 10 - 12 mm
Katalog # 229-0726
Leverandør: KARTELL
Gå til:
Product Details & Documents
PP, hvid.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Suitable for foodstuff
Med koniske ender.
Certifications: Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520.
Specifikationer
- Udv.Ø:10/11/12 mm
- Hul Ø:5,5 mm
- Rør int.Ø:10 - 12 mm
- Seq:06



