Slangekoblinger, Y-formede
6/7/8 mm, 2,7 mm, 6 - 8 mm
Katalog # 229-0724
Leverandør: KARTELL
Gå til:
Product Details & Documents
PP, hvid.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Suitable for foodstuff
Med koniske ender.
Certifications: Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520.
Specifikationer
- Udv.Ø:6/7/8 mm
- Hul Ø:2,7 mm
- Rør int.Ø:6 - 8 mm
- Seq:04
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



