Orders LinkContactUsLinkComponent
Slangekoblinger, lige
Slangekoblinger, lige
14/15/16 mm, 10,5 mm, 13-16 mm
Katalog # 229-0718
Leverandør:   KARTELL
Slangekoblinger, lige
Katalog # 229-0718
Leverandør:  KARTELL
Slangekoblinger, lige
Slangekoblinger, lige
14/15/16 mm, 10,5 mm, 13-16 mm
Price
116,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
PP, hvid
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.
Specifikationer
  • Udv.Ø:
    14/15/16 mm
  • Hul Ø:
    10,5 mm
  • Rør int.Ø:
    13-16 mm