Slangekoblinger, lige
12/13/14 mm, 8,5 mm, 11-14 mm
: KARTELL
PP, hvid
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.
- Udv.Ø:12/13/14 mm
- Hul Ø:8,5 mm
- Rør int.Ø:11-14 mm