Orders LinkContactUsLinkComponent
Slangekoblinger, lige
12/13/14 mm, 8,5 mm, 11-14 mm
  229-0717
 :   KARTELL
Slangekoblinger, lige
  229-0717
 :  KARTELL
Slangekoblinger, lige
Slangekoblinger, lige
12/13/14 mm, 8,5 mm, 11-14 mm
97,10 kr
1 * 10 Items
PP, hvid
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.
  • Udv.Ø:
    12/13/14 mm
  • Hul Ø:
    8,5 mm
  • Rør int.Ø:
    11-14 mm