Orders LinkContactUsLinkComponent
Slangekoblinger, lige
Slangekoblinger, lige
10/11/12 mm, 6,5 mm, 9-12 mm
Katalog # 229-0716
Leverandør:   KARTELL
Slangekoblinger, lige
Katalog # 229-0716
Leverandør:  KARTELL
Slangekoblinger, lige
Slangekoblinger, lige
10/11/12 mm, 6,5 mm, 9-12 mm
Price
86,30 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
PP, hvid
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.
Specifikationer
  • Udv.Ø:
    10/11/12 mm
  • Hul Ø:
    6,5 mm
  • Rør int.Ø:
    9-12 mm