Slangekoblinger, lige
6/7/8 mm, 3,5 mm, 5-7 mm
Leverandør: KARTELL
Gå til:
Product Details & Documents
PP, hvid
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.
Specifikationer
- Udv.Ø:6/7/8 mm
- Hul Ø:3,5 mm
- Rør int.Ø:5-7 mm