Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cleaning brushes for samplers

Cleaning brushes for samplers

Nylon, 15 - 30 mm, Wire, 1600 mm
  300-0493
Cleaning brushes for samplers
  300-0493
Cleaning brushes for samplers
Cleaning brushes for samplers

Nylon, 15 - 30 mm, Wire, 1600 mm

 
501,00 kr
   
1 * 1 Items
 

Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.

  • Available in a range of lengths and diameters

Documents

 
  • Børstemateriale:
    Nylon
  • Børstehoved Ø:
    15 - 30 mm
  • Handle material:
    Wire
  • Seq:
    07
  • Samlet længde:
    1600 mm