Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
300 mm, 29/32 NS
Katalog # WITG1.205.301
Leverandør:   witeg Labortechnik
Distilling bridges
Katalog # WITG1.205.301
Leverandør:  witeg Labortechnik
Distilling bridges
Distilling bridges
300 mm, 29/32 NS
Price
1.480,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kondenserlængde:
    300 mm