Distilling bridges
300 mm, 24/29 NS
Katalog # WITG1.205.300
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Specifikationer
- Konusstr.:24/29 NS
- Kondenserlængde:300 mm