Orders LinkContactUsLinkComponent
Protein G
Protein G fluorescein conjugated, 1 mg
Katalog # ROCKPG00-02
Leverandør:   Rockland Immunochemicals
Protein G
Katalog # ROCKPG00-02
CAS nummer:  
Protein G
Protein G
Protein G fluorescein conjugated, 1 mg
CAS nummer:  
Price
5.030,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents
Protein G is a surface protein of two groups of streptococcal bacteria that has ability to bind immunoglobulins and also to albumins. Protein G are prepared from chromatographically pure protein G.
Protein G is stable at 4 °C as an undiluted liquid. Freezing alkaline phosphatase conjugates will result in a substantial loss of enzymatic activity.
Protein G alkalline phosphatase conjugated is ideal for signal transduction, molecular biology, cancer, and cell signaling. Protein G peroxidase conjugated is a useful reagent in western blotting and ELISA experiments. Protein G fluorescein conjugated is suitable for immunomicroscopy and flow cytometry or FACS analysis.
Specifikationer
Specifikationer
  • Pakn:
    1 mg
  • Beskrivelse:
    Protein G fluorescein conjugated
Specifikationer