Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
5 mg, 0,2 ± mg
Katalog # 611-8230
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M1
Katalog # 611-8230
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
5 mg, 0,2 ± mg
Price
104,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Specifikationer
  • Vægt:
    5 mg
  • Seq:
    27
  • Tolerance:
    0,2 ± mg