Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
20 ml, 10/13, 0,04 ± ml
Katalog # POULRB258/D/5
Leverandør:   Poulten & Graf
Volumetric flasks, class A
Katalog # POULRB258/D/5
Leverandør:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
20 ml, 10/13, 0,04 ± ml
Price
785,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifikationer
  • NS:
    10/13
  • Kapacitet:
    20 ml
  • Seq:
    21
  • Tolerance:
    0,04 ± ml