6-Methoxyindol ≥98%
Glass bottle for solids, 250 mg
Purity: 98+% CAS No.: 3189-13-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 90-94° Boiling Point: 105°/0.2mm MDL No.: MFCD00022780
: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
- :3189-13-7
- :C₉H₉NO
- :MFCD00022780
- :147.18 g/mol
- :105 °C (0,2 mmHg)
- :90…94 °C
- :Stue temperatur
- :221-689-6
- Pakning:Glass bottle for solids
- Pakn:250 mg