Orders LinkContactUsLinkComponent
6-Methoxyindol ≥98%
Glass bottle for solids, 250 mg
  L19074.MD
6-Methoxyindol ≥98%
  L19074.MD
 :  3189-13-7
6-Methoxyindol ≥98%
6-Methoxyindol ≥98%
Glass bottle for solids, 250 mg
 :  3189-13-7
912,00 kr
1 * 250 mg
Purity: 98+% CAS No.: 3189-13-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 90-94° Boiling Point: 105°/0.2mm MDL No.: MFCD00022780
 :  6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
  •  :3189-13-7
  •  :C₉H₉NO
  •  :MFCD00022780
  •  :147.18 g/mol
  •  :105 °C (0,2 mmHg)
  •  :90…94 °C
  •  :Stue temperatur
  •  :221-689-6
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 mg