PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
Product Details & Documents
CP-PoraPLOT U and CP-PoraPLOT S provide symmetrical peaks from polar and non-polar volatiles for ultimate accuracy.
- Minimal particle loss reduces detector spiking for reliable results
- Repeatable retention times for better long-term stability
CP-PoraPLOT U provides symmetrical peaks with polar volatiles. Water has no effect on retention times and elutes as a sharp and quantifiable peak. CP-PoraPLOT U is the most polar porous polymer PLOT column and is designed for halogenated compounds, hydrocarbons C1-C6, ketones and solvents.
CP-PoraPLOT S is a divinylbenzene/vinylpyridine polymer for hydrocarbons and ketones. This phase is ideal for the analysis of medium polarity volatiles, including hydrocarbons and ketones, at higher temperatures than CP-PoraPLOT U.
Specifications
- Kolonne form:Kapillær
- Filmtykkelse:20,00 µm
- Ø indv.:0,53 mm
- Længde:10 m
- Seq:04
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



