Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Propylacetat ≥99%

Propylacetat ≥99%

Glass bottle, 1.000 mL
Katalog # L15355.AU
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Propylacetat ≥99%
Katalog # L15355.AU
CAS nummer:  109-60-4
Propylacetat ≥99%
Propylacetat ≥99%

Glass bottle, 1.000 mL

CAS nummer:  109-60-4
Price
503,00 kr
Log ind for tilgængelighed
1 * 1.000 mL
Product Details & Documents

Purity: 99% CAS No.: 109-60-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 14°(57°F) Refractive Index: 1.3840 Odour: Fruit-like Melting Point: -92° Boiling Point: 99-102° Beilstein Reference: 1740764 Merck Reference: 14,7841 UN No.: 1276 MDL No.: MFCD00009372

Synonymer:  Acetic acid propyl ester
  • CAS nummer:109-60-4
  • Formel:C₅H₁₀O₂
  • MDL nummer:MFCD00009372
  • Molecular Weight:102.13 g/mol
  • Merck Index:14,7841
  • Kogende Pt:99…102 °C
  • Smeltende Pt:–92 °C
  • Tæthed:0,887
  • Flash Pt:14 °C (57 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-686-1
  • FN:1276
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1.000 mL

Specifikationer