Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylacetat ≥99%

Methylacetat ≥99%

Glass bottle, 1.000 mL
Catalog # L14475.AU
Flammable
Irritant
Methylacetat ≥99%
Catalog # L14475.AU
CAS Number:  79-20-9
Methylacetat ≥99%
Methylacetat ≥99%

Glass bottle, 1.000 mL

CAS Number:  79-20-9
Price
419,00 kr
Log in for availability
1 * 1.000 mL
Product Details & Documents

Purity: 99% CAS No.: 79-20-9 Molecular Formula: CH3COOCH3 Formula Weight: 74.08 Flash Point: -10°(14°F) Refractive Index: 1.3620 Odour: Pleasant Melting Point: -98° Boiling Point: 56-58° Beilstein Reference: 1736662 Merck Reference: 14,6008 UN No.: 1231 MDL No.: MFCD00008711

Synonyms:  Acetic acid methyl ester
  • CAS Number:79-20-9
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00008711
  • Molecular Weight:74.08 g/mol
  • Merck Index:14,6008
  • Boiling Pt:56…58 °C
  • Melting Pt:–98 °C
  • Density:0,934
  • Flash Pt:–10 °C (14 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:201-185-2
  • UN:1231
  • ADR:3,II
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1.000 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.