Methylethylketon ≥99%
Glass bottle, 1.000 mL
Methylethylketon ≥99%
L13185.AU
: 78-93-3
Purity: 99% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Refractive Index: 1.3785 Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648
: Ethylmethylketon, Butanon
- :78-93-3
- :C₂H₅COCH₃
- :MFCD00011648
- :72.11 g/mol
- :14,6072
- :80 °C
- :–87 °C
- :0,805
- :–3 °C (26 °F)
- :Stue temperatur
- :201-159-0
- :1193
- :3,II
- :01-2119457290-43
- Pakning:Glass bottle
- Pakn:1.000 mL
- Cat. No.:L13185.AU
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



