Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3,5-Dihydroxybenzonitril ≥98%

3,5-Dihydroxybenzonitril ≥98%

Glass bottle for solids, 1 g
Katalog # L05642.03
Giftig
Irriterende
Leverandør:   Thermo Fisher Scientific
3,5-Dihydroxybenzonitril ≥98%
Katalog # L05642.03
CAS nummer:  19179-36-3
3,5-Dihydroxybenzonitril ≥98%
3,5-Dihydroxybenzonitril ≥98%

Glass bottle for solids, 1 g

CAS nummer:  19179-36-3
Price
882,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 19179-36-3 Molecular Formula: C7H5NO2 Formula Weight: 135.12 Melting Point: 185-190° Beilstein Reference: 6322491 UN No.: 3439 MDL No.: MFCD00016453

  • CAS nummer:19179-36-3
  • Formel:C₇H₅NO₂
  • MDL nummer:MFCD00016453
  • Molecular Weight:135.12 g/mol
  • Kogende Pt:311 °C (1013 hPa)
  • Smeltende Pt:185…190 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:242-859-6
  • FN:3439
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.