Orders LinkContactUsLinkComponent
Water testing kit, MINIKIT
Sulphite (high range), 20 - 500 mg/l Na2SO3, AF 435, 1 KIT
Katalog # 705-0421
Leverandør:   Lovibond
Water testing kit, MINIKIT
Katalog # 705-0421
Leverandør:  Lovibond
CAS nummer:  
Water testing kit, MINIKIT
Water testing kit, MINIKIT
Sulphite (high range), 20 - 500 mg/l Na2SO3, AF 435, 1 KIT
CAS nummer:  
Price
719,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.
  • MINIKITS are based on titrimetric methods
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years
Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.
Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Sulphite (high range)
  • Model:
    AF 435
  • Område:
    20 - 500 mg/l Na2SO<sub>3</sub>
Specifikationer
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 100 ml
    Dilution/shaker tube with lid, plastic, graduated, 100 ml
    LOVI385130
    Log ind for tilgængelighed
    1 * 1 Items - 112,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 30 ml
    Dilution/shaker tube with lid, plastic, graduated, 30 ml
    LOVI385132
    Log ind for tilgængelighed
    1 * 1 Items - 112,00 kr